Learn More
Medchemexpress LLC Urea, N-[4-(1-amino-5-isoquinolinyl)phenyl]-N'-[5-(1,1-dimethylethyl)-3-isoxazolyl]- | 1481641-08-0 | 98.9% | 401.46 | 50 MG
SDP
Catalog No. 5000385494
Encompass_Preferred
RIPK1-IN-4 is a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase. It binds to a DLG-out inactive form of RIP1 with IC50s of 16 nM for RIP1 and 10 nM for ADP-Glo kinase.
  • Potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase.
  • Binds to a DLG-out inactive form of RIP1.
  • For research use only.
Catalog No. 50-003-85494 Supplier Medchemexpress LLC Supplier No. HY1890150MG
RIPK1-IN-4 is a potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase. It binds to a DLG-out inactive form of RIP1 with IC50s of 16 nM for RIP1 and 10 nM for ADP-Glo kinase.
  • Potent and selective type II kinase inhibitor of receptor interacting protein 1 (RIP1) kinase.
  • Binds to a DLG-out inactive form of RIP1.
  • For research use only.