Learn More
Medchemexpress LLC Urea, N-[4-[4-(4-morpholinyl)-6-(3-oxa-8-azabicyclo[3.2.1]oct-8-yl)-1,3,5-triazin-2-yl]phenyl]-N'-4-pyridinyl-, hydrochloride (1:1) | 1463510-35-1 | 99.7% | 525.00 | 25 MG
SDP
Catalog No. 5000345640
Encompass_Preferred
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It shows anti-tumor activity in vivo, inhibiting tumor growth and PI3K signaling without significant weight loss in animal models. The product is suitable for laboratory research applications.
  • Potent and orally active dual PI3K/mTOR inhibitor
  • Anti-tumor activity in vivo
  • Inhibits cell proliferation
  • Available as a white to off-white solid
  • Tested and complies with specifications
Catalog No. 50-003-45640 Supplier Medchemexpress LLC Supplier No. 25MGHY11080A
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It shows anti-tumor activity in vivo, inhibiting tumor growth and PI3K signaling without significant weight loss in animal models. The product is suitable for laboratory research applications.
  • Potent and orally active dual PI3K/mTOR inhibitor
  • Anti-tumor activity in vivo
  • Inhibits cell proliferation
  • Available as a white to off-white solid
  • Tested and complies with specifications