Learn More
Medchemexpress LLC Urea, N-[4-[8-amino-3-(1-methylethyl)imidazo[1,5-a]pyrazin-1-yl]-1-naphthalenyl]-N'-[3-(trifluoromethyl)phenyl]- | 1414938-21-8 | 99.2% | 504.51 | 5 MG
SDP
Catalog No. 5000387909
Encompass_Preferred
IRE1α kinase-IN-2 is a potent IRE1α kinase inhibitor with an EC50 of 0.82 μM. It inhibits IRE1α kinase autophosphorylation (IC50=3.12 μM) and XBP1 mRNA splicing in WT cell lines.
  • Potent IRE1α kinase inhibitor.
  • Inhibits IRE1α kinase autophosphorylation.
  • Inhibits XBP1 mRNA splicing.
  • For research use only.
Catalog No. 50-003-87909 Supplier Medchemexpress LLC Supplier No. HY185095MG
IRE1α kinase-IN-2 is a potent IRE1α kinase inhibitor with an EC50 of 0.82 μM. It inhibits IRE1α kinase autophosphorylation (IC50=3.12 μM) and XBP1 mRNA splicing in WT cell lines.
  • Potent IRE1α kinase inhibitor.
  • Inhibits IRE1α kinase autophosphorylation.
  • Inhibits XBP1 mRNA splicing.
  • For research use only.