Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urea, N-[4-chloro-2-hydroxy-3-(3-methyltetrahydrofuran-3-yl)sulfonylphenyl]-N'-(2-methylcyclopentenyl) | 1838123-22-0 | 99.4% | 50 MG
SDP

Catalog No. 5000232620
Encompass_Preferred

(R,R)-CXCR2-IN-2 is the (R,R) diastereoisomer of CXCR2-IN-2, a selective, brain-penetrant CXCR2 antagonist used as a research tool in preclinical studies. It is supplied as a high-purity solid for laboratory use.

  • Brain-penetrant CXCR2 antagonist active in cell-based assays.
  • High purity (reported >99%).
  • Molecular formula C18H23ClN2O5S; molecular weight 414.90 g/mol.
  • Supplied as a solid suitable for dissolution in DMSO for in vitro assays.
  • Intended for research use only; not for human use.

Catalog No. 50-002-32620 Supplier Medchemexpress LLC Supplier No. HY120878A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R,R)-CXCR2-IN-2 is the (R,R) diastereoisomer of CXCR2-IN-2, a selective, brain-penetrant CXCR2 antagonist used as a research tool in preclinical studies. It is supplied as a high-purity solid for laboratory use.

  • Brain-penetrant CXCR2 antagonist active in cell-based assays.
  • High purity (reported >99%).
  • Molecular formula C18H23ClN2O5S; molecular weight 414.90 g/mol.
  • Supplied as a solid suitable for dissolution in DMSO for in vitro assays.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.