Learn More
Medchemexpress LLC Urea, N-[[4-methyl-2-(2-thienyl)-5-thiazolyl]methyl]-N'-2-thienyl- | 1396814-79-1 | 98.5% | 335.47 | 25 MG
SDP
Catalog No. 5000394596
Encompass_Preferred
VU0542270 is a selective vascular Kir6.1/SUR2B KATP channel inhibitor with an IC50 value of 100 nM. It has an IC50 > 30 μM for the other nine members of the Kir channel family. It can be used in the study of cardiovascular disease.
  • Selective vascular Kir6.1/SUR2B KATP channel inhibitor.
  • Can be used in the study of cardiovascular disease.
  • High selectivity, with IC50 > 30 μM for other Kir channel family members.
Catalog No. 50-003-94596 Supplier Medchemexpress LLC Supplier No. 25MGHY162322
VU0542270 is a selective vascular Kir6.1/SUR2B KATP channel inhibitor with an IC50 value of 100 nM. It has an IC50 > 30 μM for the other nine members of the Kir channel family. It can be used in the study of cardiovascular disease.
  • Selective vascular Kir6.1/SUR2B KATP channel inhibitor.
  • Can be used in the study of cardiovascular disease.
  • High selectivity, with IC50 > 30 μM for other Kir channel family members.