Learn More
Medchemexpress LLC Urea, N'-cyclohexyl-N-(phenylmethyl)-N-(4-piperidinylmethyl) hydrochloride | 2070014-88-7 | MFCD30377213 | 99.6% | 365.94 | C20H32ClN3O | 100 MG
SDP
Catalog No. 5000214397
Encompass_Preferred
SRI-011381 hydrochloride is a research compound that functions as an orally active agonist of TGF-β/Smad signaling and has demonstrated neuroprotective effects in preclinical studies. It is supplied as a white to off-white solid and is characterized for both in vitro and in vivo research use.
  • Orally active TGF-β signaling agonist with reported neuroprotective effects.
  • High purity (~99.6%) suitable for research applications.
  • White to off-white solid with validated physicochemical data.
  • Soluble in DMSO and water; multiple in vivo formulation protocols available.
  • Supplied in multiple sizes for dosing flexibility, including 100 mg.
Catalog No. 50-002-14397 Supplier Medchemexpress LLC Supplier No. HY100347A100MG
SRI-011381 hydrochloride is a research compound that functions as an orally active agonist of TGF-β/Smad signaling and has demonstrated neuroprotective effects in preclinical studies. It is supplied as a white to off-white solid and is characterized for both in vitro and in vivo research use.
  • Orally active TGF-β signaling agonist with reported neuroprotective effects.
  • High purity (~99.6%) suitable for research applications.
  • White to off-white solid with validated physicochemical data.
  • Soluble in DMSO and water; multiple in vivo formulation protocols available.
  • Supplied in multiple sizes for dosing flexibility, including 100 mg.