Learn More
Medchemexpress LLC Urea, N'-methyl-N,N-diphenyl- | 13114-72-2 | 226.28 | 100 G
SDP
Catalog No. 5000447645
Encompass_Preferred
Akardite II (AK-II) is a stabilizer for nitrocellulose (NC) propellants by scavenging NOx. It also helps control various plant pathogens and weeds, functioning as a fungicide and herbicide. This product is for research use only.
  • Stabilizer for nitrocellulose (NC) propellants
  • Scavenges NOx
  • Helps control plant pathogens
  • Functions as a fungicide and herbicide
  • For research use only
Catalog No. 50-004-47645 Supplier Medchemexpress LLC Supplier No. HYW004643100G
Akardite II (AK-II) is a stabilizer for nitrocellulose (NC) propellants by scavenging NOx. It also helps control various plant pathogens and weeds, functioning as a fungicide and herbicide. This product is for research use only.
  • Stabilizer for nitrocellulose (NC) propellants
  • Scavenges NOx
  • Helps control plant pathogens
  • Functions as a fungicide and herbicide
  • For research use only