Learn More
Medchemexpress LLC Urea, N,N'-dicyclohexyl | 2387-23-7 | MFCD00003829 | ≥95.0% | 224.34 g/mol | C13H24N2O | 50 MG
SDP
Catalog No. 5000304000
Encompass_Preferred
1,3-Dicyclohexylurea is a small-molecule research compound that functions as a potent soluble epoxide hydrolase (sEH) inhibitor. It is provided as a white to off-white solid suitable for analytical standards, formulation studies, and pharmacology research, with batch-specific certificates of analysis documenting identity and purity.
  • Potent sEH inhibitor for biochemical and pharmacology research.
  • High purity (≥95.0%) suitable for analytical and in vivo studies.
  • Molecular weight 224.34 g/mol and formula C13H24N2O.
  • White to off-white solid with defined storage recommendations for stability.
  • Available in small research pack sizes for formulation and assay development.
  • Certificate of analysis available for batch-specific quality verification.
Catalog No. 50-003-04000 Supplier Medchemexpress LLC Supplier No. HYW01398950MG
1,3-Dicyclohexylurea is a small-molecule research compound that functions as a potent soluble epoxide hydrolase (sEH) inhibitor. It is provided as a white to off-white solid suitable for analytical standards, formulation studies, and pharmacology research, with batch-specific certificates of analysis documenting identity and purity.
  • Potent sEH inhibitor for biochemical and pharmacology research.
  • High purity (≥95.0%) suitable for analytical and in vivo studies.
  • Molecular weight 224.34 g/mol and formula C13H24N2O.
  • White to off-white solid with defined storage recommendations for stability.
  • Available in small research pack sizes for formulation and assay development.
  • Certificate of analysis available for batch-specific quality verification.