Learn More
Medchemexpress LLC Urea, N,N'-dicyclohexyl- | 2387-23-7 | MFCD00003829 | 99.9% | 224.34 g/mol | C13H24N2O | 10 MG
SDP
Catalog No. 5000303811
Encompass_Preferred
1,3-Dicyclohexylurea is a small-molecule research chemical used as a soluble epoxide hydrolase (sEH) inhibitor for biochemical and pharmacological studies. It is supplied as a high-purity analytical standard suitable for assay development, inhibitor screening, and analytical reference. Manufacturer documentation typically includes a certificate of analysis and a safety data sheet for traceability and safe handling.
  • High purity suitable for analytical applications.
  • Reported purity 99.85% by certificate of analysis.
  • Molecular formula C13H24N2O and molecular weight 224.34 g/mol.
  • Used as a soluble epoxide hydrolase inhibitor in biochemical studies.
  • Available in small and larger package sizes to support assay development.
  • Accompanied by COA and SDS for quality and safety information.
Catalog No. 50-003-03811 Supplier Medchemexpress LLC Supplier No. HYW013989R10MG
1,3-Dicyclohexylurea is a small-molecule research chemical used as a soluble epoxide hydrolase (sEH) inhibitor for biochemical and pharmacological studies. It is supplied as a high-purity analytical standard suitable for assay development, inhibitor screening, and analytical reference. Manufacturer documentation typically includes a certificate of analysis and a safety data sheet for traceability and safe handling.
  • High purity suitable for analytical applications.
  • Reported purity 99.85% by certificate of analysis.
  • Molecular formula C13H24N2O and molecular weight 224.34 g/mol.
  • Used as a soluble epoxide hydrolase inhibitor in biochemical studies.
  • Available in small and larger package sizes to support assay development.
  • Accompanied by COA and SDS for quality and safety information.