Learn More
Medchemexpress LLC Urea, N,N'-dicyclohexyl- | 2387-23-7 | MFCD00003829 | 99.9% | 224.34 g/mol | C13H24N2O | 100 MG
SDP
Catalog No. 5000304049
Encompass_Preferred
1,3-Dicyclohexylurea is a small-molecule soluble epoxide hydrolase (sEH) inhibitor supplied as a research-grade analytical standard (CAS 2387-23-7). It has the molecular formula C13H24N2O and molecular weight 224.34 g/mol, and is accompanied by analytical documentation for laboratory use.
  • High purity (99.85% by HPLC) for reliable analytical results.
  • Available in multiple pack sizes from milligram to gram scales.
  • Provided with certificate of analysis, SDS, and analytical data.
  • Supplied as solid or solution formats for flexible use.
  • Suitable as an analytical reference and for enzyme inhibition studies.
  • Packaged in brown glass bottle to protect from light and contamination.
Catalog No. 50-003-04049 Supplier Medchemexpress LLC Supplier No. HYW013989R100MG
1,3-Dicyclohexylurea is a small-molecule soluble epoxide hydrolase (sEH) inhibitor supplied as a research-grade analytical standard (CAS 2387-23-7). It has the molecular formula C13H24N2O and molecular weight 224.34 g/mol, and is accompanied by analytical documentation for laboratory use.
  • High purity (99.85% by HPLC) for reliable analytical results.
  • Available in multiple pack sizes from milligram to gram scales.
  • Provided with certificate of analysis, SDS, and analytical data.
  • Supplied as solid or solution formats for flexible use.
  • Suitable as an analytical reference and for enzyme inhibition studies.
  • Packaged in brown glass bottle to protect from light and contamination.