Learn More
Medchemexpress LLC Urea, N,N'-dicyclohexyl- | 2387-23-7 | MFCD00003829 | 99.9% | 224.34 g/mol | C13H24N2O | 5 MG
SDP
Catalog No. 5000304171
Encompass_Preferred
1,3-Dicyclohexylurea is a research-grade small molecule described as an orally active, potent soluble epoxide hydrolase (sEH) inhibitor used in biochemical and pharmacological studies. It is provided as a solid for laboratory research and has been used in both in vitro and in vivo experiments.
Catalog No. 50-003-04171 Supplier Medchemexpress LLC Supplier No. HYW0139895MG
1,3-Dicyclohexylurea is a research-grade small molecule described as an orally active, potent soluble epoxide hydrolase (sEH) inhibitor used in biochemical and pharmacological studies. It is provided as a solid for laboratory research and has been used in both in vitro and in vivo experiments.