Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ Urea Reagents

Catalog No. 01818134 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
R1 4 x 165 mL, R2 4 x 165 mL
R1 4 x 53 mL, R2 4 x 53 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
01-818-134 R1 4 x 53 mL, R2 4 x 53 mL
01-801-020 R1 4 x 165 mL, R2 4 x 165 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-818-134 Supplier Beckman Coulter™ Supplier No. OSR6234
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Urea R1 and R2 for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Includes R1 4 x 53 mL, R2 4 x 53 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.