Learn More
U.S. Pharmacopeia Urea Related Compound A, 108-19-0, MFCD00007946, 15 mg
Catalog No. NC1166110
Encompass
Linear Formula: NH2CONHCONH2, Molecular Weight: 103.08, mp: 185-190 °C, storage temp: 2-8°C, Synonym: Allophanic acid amide, Biuret, Carbamoylurea.
Catalog No. NC1166110 Supplier U.S. Pharmacopeia Supplier No. 1706712
Linear Formula: NH2CONHCONH2, Molecular Weight: 103.08, mp: 185-190 °C, storage temp: 2-8°C, Synonym: Allophanic acid amide, Biuret, Carbamoylurea.