Learn More
U.S. Pharmacopeia Urea, 57-13-6, MFCD00008022, 200mg
Catalog No. NC0163226
Encompass
Molecular formula CH4N2O, Molecular weight 60.06, Melting point 269.6 - 275 °F (132 - 135 °C), Boiling point 275 °F (135 °C) (decomposes), Synonyms: Carbamide, Carbonyl diamide
Catalog No. NC0163226 Supplier U.S. Pharmacopeia Supplier No. 1706698
Molecular formula CH4N2O, Molecular weight 60.06, Melting point 269.6 - 275 °F (132 - 135 °C), Boiling point 275 °F (135 °C) (decomposes), Synonyms: Carbamide, Carbonyl diamide