Learn More
National Diagnostics UREA - ULTA PURE 1 KG
SDP
Supplier:  National Diagnostics EC6051KG
Encompass
Urea - ULTA PURE, Recrystallized to remove ammonia. Exclusive purification process, 1 KG
Catalog No. 50-899-90046