Learn More
Alkali Scientific Urea, UltraPure (USP Grade), 2.5 Kg
SDP
Catalog No. 5000660572
Encompass_Preferred
Urea UltraPure (USP Grade) is available in a 2.5kg quantity and is used widely in laboratory settings for various biochemical applications. Urea serves as an essential nitrogen source in protein synthesis and is commonly used in buffer preparations and other analytical procedures. This ultra-pure grade ensures the highest level of purity, making it suitable for applications in clinical, academic, and industrial research. Urea is used in studies of protein denaturation, enzyme activity, and in the formulation of various laboratory solutions. The 2.5kg size is perfect for medium-to-large scale laboratory use and ensures that researchers have a reliable supply for their experiments.
Catalog No. 50-006-60572 Supplier Alkali Scientific Supplier No. URE003CP
Urea UltraPure (USP Grade) is available in a 2.5kg quantity and is used widely in laboratory settings for various biochemical applications. Urea serves as an essential nitrogen source in protein synthesis and is commonly used in buffer preparations and other analytical procedures. This ultra-pure grade ensures the highest level of purity, making it suitable for applications in clinical, academic, and industrial research. Urea is used in studies of protein denaturation, enzyme activity, and in the formulation of various laboratory solutions. The 2.5kg size is perfect for medium-to-large scale laboratory use and ensures that researchers have a reliable supply for their experiments.