Learn More
Research Products International Corp Replacement Urea Vial Caps, 22mm Polyseal Cone Lined Plastic Screw Caps, White, 1000 per Package
SDP
Catalog No. 50213011
Encompass
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Urea Vial Caps, 22 mm Polyseal Cone Lined, White, Plastic Screw Cap
Catalog No. 50-213-011 Supplier Research Products International Corp Supplier No. 131002
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Urea Vial Caps, 22 mm Polyseal Cone Lined, White, Plastic Screw Cap