Learn More
Research Products International Corp Replacement Urea Vial Caps, 22mm Poly Disc Lined Plastic Screw Caps, White, 1000 per Package
SDP
Catalog No. 50213010
Encompass
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Urea Vial Caps, 22 mm Poly Disc Lined Plastic Screw Caps, White
Catalog No. 50-213-010 Supplier Research Products International Corp Supplier No. 131001
Smooth top white caps available with various type liners for liquid scintillation counting applications. Recommended for all samples that have a pH of 5-7. For highly alkaline or acidic samples an inert liner is recommended.
Urea Vial Caps, 22 mm Poly Disc Lined Plastic Screw Caps, White