Learn More
Enzo Life Sciences AMPIPROBE Ureaplasma spp. / M. genitalium / M. hominis Assay Kit (100tests)
SDP
Catalog No. 502013767
Encompass_Preferred
The AMPIPROBE Ureaplasma spp. / M. genitalium / M. hominis (UMM) Assay is a real-time polymerase chain reaction (qPCR) assay for the qualitative detection of Ureaplasma spp. (Ureaplasma parvum and Ureaplasma urealyticum), Mycoplasma genitalium, and Mycoplasma hominis DNA. The kit uses the AMPIPROBE assay platform which takes advantage of paired fluorophore- and quencher-labeled primers specific for each of the target species as well as an internal control. The kit contains all reagents necessary for PCR-based detection of Ureaplasma spp., Mycoplasma genitalium, and Mycoplasma hominis DNA. A positive PCR control consisting of a mixture of the target templates and a negative PCR control that results negative for UMM, but positive for the internal control (human β-globin). Please read the complete kit insert before performing this assay. Applications: qPCR. Species reactivity: Mycoplasma hominis, Mycoplasma genitalium, Ureaplasma spp.
Catalog No. 50-201-3767 Supplier Enzo Life Sciences Supplier No. ENZGEN2090100
The AMPIPROBE Ureaplasma spp. / M. genitalium / M. hominis (UMM) Assay is a real-time polymerase chain reaction (qPCR) assay for the qualitative detection of Ureaplasma spp. (Ureaplasma parvum and Ureaplasma urealyticum), Mycoplasma genitalium, and Mycoplasma hominis DNA. The kit uses the AMPIPROBE assay platform which takes advantage of paired fluorophore- and quencher-labeled primers specific for each of the target species as well as an internal control. The kit contains all reagents necessary for PCR-based detection of Ureaplasma spp., Mycoplasma genitalium, and Mycoplasma hominis DNA. A positive PCR control consisting of a mixture of the target templates and a negative PCR control that results negative for UMM, but positive for the internal control (human β-globin). Please read the complete kit insert before performing this assay. Applications: qPCR. Species reactivity: Mycoplasma hominis, Mycoplasma genitalium, Ureaplasma spp.