Learn More
ATCC Ureaplasma parvum
Supplier:  ATCC 27813
Encompass
Ureaplasma parvum strain 7 was isolated from a case of nongonococcal urethritis. This strain is propagated in B-broth medium for Ureaplasma and requires an additional 5-10% horse serum added to the broth for growth.
Catalog No. 50-238-2947