Learn More
ATCC Ureaplasma parvum
Supplier:  ATCC 27815
Encompass
Ureaplasma parvum strain 27 was isolated from a case of nongonococcal urethritis. This strain grows in B-broth medium for Ureaplasma and requires an additional 5-10% horse serum added to the broth for growth.
Catalog No. 50-238-4367