Learn More
Bioss Ureaplasma sp.
SDP
Catalog No. 5000066788
Encompass_Preferred
Ureaplasma sp Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66788 Supplier Bioss Supplier No. BSM72458M
Ureaplasma sp Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide