Learn More
ATCC Ureaplasma urealyticum
Catalog No. 502383985
Encompass
Ureaplasma urealyticum strain Western is a genome-sequenced bacterial strain. This strain requires an additional 5-10% horse serum to be added to the broth for growth.
Catalog No. 50-238-3985 Supplier ATCC Supplier No. 33699
Ureaplasma urealyticum strain Western is a genome-sequenced bacterial strain. This strain requires an additional 5-10% horse serum to be added to the broth for growth.