Learn More
Sigma Aldrich Fine Chemicals Biosciences Urease from Canavalia ensiformis
Catalog No. NC0518902
Encompass
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL, CAS Number 9002-13-5, MDL number MFCD00070858
Catalog No. NC0518902 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U1875100ML
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL, CAS Number 9002-13-5, MDL number MFCD00070858