Learn More
Sigma Aldrich Fine Chemicals Biosciences Urease Buffer Reagent
Catalog No. 502900358
Encompass
Urease is a 545 kDa protein critical for the hydrolysis of urea to carbon dioxide and ammonia.
Catalog No. 50-290-0358 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. U33831VL
Urease is a 545 kDa protein critical for the hydrolysis of urea to carbon dioxide and ammonia.