Learn More
Crescent Chemical Co Inc UREASE FROM JACK BEAN 220U/MG
SDP
Catalog No. 50248272
Encompass_Preferred
Urease from jack bean min. 220 U/mg lyophil. For the determination of urea (1). Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 37799.01 CAS 9002-13-5
Catalog No. 50-248-272 Supplier Crescent Chemical Co Inc Supplier No. SE37799.01
Urease from jack bean min. 220 U/mg lyophil. For the determination of urea (1). Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 37799.01 CAS 9002-13-5