Learn More
Cayman Chemical Urease HelIcobacTR pylorI Ra
SDP
Catalog No. 502946986
Encompass_Preferred
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) H. pylori urease • Applications: ELISA
Catalog No. 50-294-6986 Supplier Cayman Chemical Supplier No. 32343100 UG
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) H. pylori urease • Applications: ELISA