Learn More
Medchemexpress LLC Urease, jack bean | 9002-13-5 | MFCD00070858 | 545 kDa | 20 KU
SDP
Catalog No. 5000400291
Encompass_Preferred
Urease, jack bean, is an enzyme derived from jack bean that catalyzes the hydrolysis of urea into carbon dioxide and ammonia. It is utilized in the determination of urea in body fluids.
  • Catalyzes the hydrolysis of urea into carbon dioxide and ammonia.
  • Useful in the determination of urea in body fluids.
  • Has an isoelectric point of 5.0-5.2.
  • Optimal pH is 7.4.
  • Soluble in 0.2 M sodium phosphate (pH 7.0, 10 mg/mL) or pure water/other buffers.
  • MES, HEPES, and CHES buffers do not inhibit activity.
  • For research use only.
Catalog No. 50-004-00291 Supplier Medchemexpress LLC Supplier No. HYP276720KU
Urease, jack bean, is an enzyme derived from jack bean that catalyzes the hydrolysis of urea into carbon dioxide and ammonia. It is utilized in the determination of urea in body fluids.
  • Catalyzes the hydrolysis of urea into carbon dioxide and ammonia.
  • Useful in the determination of urea in body fluids.
  • Has an isoelectric point of 5.0-5.2.
  • Optimal pH is 7.4.
  • Soluble in 0.2 M sodium phosphate (pH 7.0, 10 mg/mL) or pure water/other buffers.
  • MES, HEPES, and CHES buffers do not inhibit activity.
  • For research use only.