Learn More
Bioworld UREASE (RECOMBINANT)
Supplier:  Bioworld 220607441
Encompass_Preferred
Recombinant Urease Source:E.Coli Physical Appearance: Sterile Filtered White lyophilized (freeze-dried) powder. Formulation: Each mg of protein contains 345 g Potassium Phosphate and 25 g EDTA Na2. Solubility: It is recommended to...
Catalog No. 50-253-845