Learn More
Medchemexpress LLC URELUMAB 1MG
SDP
Supplier:  Medchemexpress LLC HYP990551MG
Encompass_Preferred
Urelumab, a fully human, non-ligand binding, CD137 agonist IgG4 monoclonal antibody, enhances T-cell and natural killer-cell antitumor activity, and may enhance cytotoxic activity of Rituximab. Urelumab can be used for the research of diffuse large B-cell lymphoma (DLBCL), follicular lymphoma (FL), and other types of non-Hodgkin lymphoma (NHL).
Catalog No. NC2319378