Learn More
Medchemexpress LLC Urethane (Standard) | 51-79-6 | 99.98% | 250 MG
SDP
Catalog No. 5000414354
Encompass_Preferred
Urethane (Standard) is the analytical standard of Urethane, intended for research and analytical applications. Also known as Ethyl carbamate, it is an ethyl ester of carbamic acid and a byproduct of fermentation found in various food products. This compound can suppress bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro.
  • Analytical standard for research and analysis
  • Used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Suppresses bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro
Catalog No. 50-004-14354 Supplier Medchemexpress LLC Supplier No. 250MGHYB1207R
Urethane (Standard) is the analytical standard of Urethane, intended for research and analytical applications. Also known as Ethyl carbamate, it is an ethyl ester of carbamic acid and a byproduct of fermentation found in various food products. This compound can suppress bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro.
  • Analytical standard for research and analysis
  • Used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Suppresses bacterial, protozoal, sea urchin egg, and plant tissue growth in vitro