Learn More
Aviva Systems Biology Corp Urgcp Antibody - N-terminal region (ARP56287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Urgcp. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507, RP23-385C16.7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-137