Learn More
Pharmacal Research Laboratories Uri-Solv, 5 Gal cube
SDP
Catalog No. NC0744004
Encompass
Uri-Solv is a heavy duty concentrated phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils.
Catalog No. NC0744004 Supplier Pharmacal Research Laboratories Supplier No. 08425F
Uri-Solv is a heavy duty concentrated phosphoric acid washing detergent formulated with inhibitors that minimize corrosion. This product contains non-ionic surfactants to increase the efficiency of removing urine scale deposits and organic soils.