Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uric acid-1,3-15N2 | 62948-75-8 | MFCD00145511 | 99.6% | 170.10 | C5H4N4O3 (1,3-15N2) | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB2130S110MG

Encompass_Preferred

Uric acid-1,3-15N2 is a 15N-labeled uric acid supplied as a high-purity solid for use as an isotopically labeled internal standard in analytical and biochemical workflows. It is intended for quantitation, tracer studies, and method development in mass spectrometry and related techniques.

  • Purity 99.6% for analytical accuracy.
  • Includes 10 mg of solid material.
  • Molecular weight 170.10 for mass spectrometric reference.
  • 1,3-15N2 labeling enables tracer and quantitative studies.
  • White to off-white solid appearance, suitable for standard analytical handling.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-97520


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.