Learn More
Medchemexpress LLC Uric acid-15N2 | 62948-75-8 | 99.9% | 5 MG
SDP
Catalog No. 5000411490
Encompass_Preferred
Uric acid-15N2 is the 15N labeled Uric acid. Uric acid is an important antioxidant and scavenger of oxygen radicals, contributing to the stability of blood pressure and antioxidant stress. It removes reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, which helps inhibit lipid peroxidation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Stable heavy isotopes have been incorporated into molecules for quantitation during the development process.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of substances.
Catalog No. 50-004-11490 Supplier Medchemexpress LLC Supplier No. HYB2130S15MG
Uric acid-15N2 is the 15N labeled Uric acid. Uric acid is an important antioxidant and scavenger of oxygen radicals, contributing to the stability of blood pressure and antioxidant stress. It removes reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, which helps inhibit lipid peroxidation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Stable heavy isotopes have been incorporated into molecules for quantitation during the development process.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of substances.