Learn More
Medchemexpress LLC Uric acid-15N2 | 62948-75-8 | 99.95% | 1 MG
SDP
Catalog No. 5000409893
Encompass_Preferred
Uric acid-15N2 is the 15N labeled Uric acid. Uric acid is a scavenger of oxygen radicals and a crucial antioxidant that helps maintain blood pressure stability and antioxidant stress. It can remove reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, thereby inhibiting lipid peroxidation. This compound is intended for research use only.
  • Can be used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Helps maintain blood pressure stability.
  • Removes reactive oxygen species (ROS).
  • Inhibits lipid peroxidation.
Catalog No. 50-004-09893 Supplier Medchemexpress LLC Supplier No. HYB2130S11MG
Uric acid-15N2 is the 15N labeled Uric acid. Uric acid is a scavenger of oxygen radicals and a crucial antioxidant that helps maintain blood pressure stability and antioxidant stress. It can remove reactive oxygen species (ROS) such as singlet oxygen and peroxynitrite, thereby inhibiting lipid peroxidation. This compound is intended for research use only.
  • Can be used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Helps maintain blood pressure stability.
  • Removes reactive oxygen species (ROS).
  • Inhibits lipid peroxidation.