Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uric acid, 99%, Thermo Scientific Chemicals

Catalog No. AAA1334636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1334636 500 g
AAA1334614 25 g
AAA1334622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1334636 Supplier Thermo Scientific Chemicals Supplier No. A1334636
  • $662.50
    $516.65 / Each of 1

    Save $145.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Uric acid acts as a strong reducing agent (electron donors) and potent antioxidant. It is used as a precursor in cell culture systems and in cosmetics. Further, it is useful for serum-free medium supplement. In addition to this, it is used as a reagent in the determination of uricase and tungstate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uric acid acts as a strong reducing agent (electron donors) and potent antioxidant. It is used as a precursor in cell culture systems and in cosmetics. Further, it is useful for serum-free medium supplement. In addition to this, it is used as a reagent in the determination of uricase and tungstate.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water. Insoluble in ether and alcohol.

Notes
Stable. Incompatible with other materials such as acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-93-2
Molecular Formula C5H4N4O3
Molecular Weight (g/mol) 168.112
MDL Number MFCD00005712
InChI Key LEHOTFFKMJEONL-UHFFFAOYSA-N
Synonym uric acid, urate, 2,6,8-trioxypurine, lithic acid, 2,6,8-trihydroxypurine, 2,6,8-trioxopurine, 1h-purine-2,6,8-triol, 8-hydroxyxanthine, 7,9-dihydro-1h-purine-2,6,8 3h-trione, 1h-purine-2,6,8 3h-trione, 7,9-dihydro
PubChem CID 1175
ChEBI CHEBI:17775
IUPAC Name 7,9-dihydro-3H-purine-2,6,8-trione
SMILES C12=C(NC(=O)N1)NC(=O)NC2=O

Specifications

Melting Point >300°C
Density 1.89
Quantity 500 g
Beilstein 156158
Merck Index 14,9875
Solubility Information Soluble in 1M sodium hydroxide solution. Slightly soluble in water. Insoluble in ether and alcohol.
Formula Weight 168.11
Percent Purity 99%
Chemical Name or Material Uric acid
EINECSNumber 200-720-7
RTECSNumber YU7050080
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.