Learn More
Beckman Coulter Uric Acid Determinant
Catalog No. 21280060 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
68 mL
Catalog No. Quantity
21-280-060 68 mL
1 options
This item is not returnable. View return policy
Catalog No. 21-280-060 Supplier Beckman Coulter Supplier No. OSR6098
Add to Cart
Add to Cart
This item is not returnable. View return policy
Uric Acid assay offers exceptional clinical quality for General Chemistry testing.
  • Our broad and growing portfolio of high-performing Six Sigma assays is compatible with AU Chemistry Analyzers - and it offers exceptional clinical quality
  • Load it and leave it with liquid ready-to-use reagents
  • Trust results with standardized test menus, assay protocols and reference ranges providing commutable patient results across AU analyzers
TRUSTED_SUSTAINABILITY
Specifications
Type Reagent
For Use With (Equipment) Beckman Coulter Analyzer
Quantity 68 mL