Learn More
MOLECULAR DEPOT LLC Uric Acid Interference - 1 mL
Catalog No. 5000013364
Encompass_Preferred
Uric Acid for Interference Testing consists of a highly concentrated solution of Uric Acid (20 mg/mL) provided with the corresponding diluent (diluent C) This solution can be spiked into specimens to test Uric Acid interference and tolerance in assays For in vitro research use only
Catalog No. 50-000-13364 Supplier MOLECULAR DEPOT LLC Supplier No. B2010199
Uric Acid for Interference Testing consists of a highly concentrated solution of Uric Acid (20 mg/mL) provided with the corresponding diluent (diluent C) This solution can be spiked into specimens to test Uric Acid interference and tolerance in assays For in vitro research use only