Learn More
Cayman Chemical UrIc AcIdodIum salt 2mg
SDP
Catalog No. 502970554
Encompass_Preferred
An end product of purine metabolism in humans that is mainly excreted in urine and can act as a potent peroxynitrite scavenger and antioxidant; high levels of serum uric acid (>120 UG/ml) are associated with gout, kidney stones, metabolic syndrome, hypertension, renal disease, and cardiovascular disease
Catalog No. 50-297-0554 Supplier Cayman Chemical Supplier No. 162192 MG
Add to Cart
Add to Cart
An end product of purine metabolism in humans that is mainly excreted in urine and can act as a potent peroxynitrite scavenger and antioxidant; high levels of serum uric acid (>120 UG/ml) are associated with gout, kidney stones, metabolic syndrome, hypertension, renal disease, and cardiovascular disease