Learn More
Pharmacal Research Laboratories Urid, 30 Gal
SDP
Catalog No. NC0723544
Encompass
Urid is a unique citric acid detergent that strips urine scale quickly and efficiently from cages and trays, floors and walls. This product is safer to use because it contains no strong, corrosive mineral acids. Urid penetrates, dissolves and releases the mineral bonding agents that cause adherent scale. Urid is recommended for manual use as well as in mechanical cage washers.
Catalog No. NC0723544 Supplier Pharmacal Research Laboratories Supplier No. 83030F
Urid is a unique citric acid detergent that strips urine scale quickly and efficiently from cages and trays, floors and walls. This product is safer to use because it contains no strong, corrosive mineral acids. Urid penetrates, dissolves and releases the mineral bonding agents that cause adherent scale. Urid is recommended for manual use as well as in mechanical cage washers.