Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine-1',2',3',4',5'-13C5 | 159496-16-9 | 99.9% | 249.16 g/mol | C4(13C)5H12N2O6 | 10 MG
SDP

Catalog No. 5000297268
Encompass_Preferred

Uridine-13C5 is a stable isotope-labeled form of the nucleoside uridine with five 13C-enriched carbon atoms. It is supplied as a research-grade solid intended for use as an internal standard or tracer in metabolic labeling and mass spectrometry applications.

  • Purity 99.9%.
  • Physical form off-white to light brown solid.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for use as an internal standard or tracer in metabolic and mass spectrometry studies.
  • Available in small packaging sizes, including 10 mg.
  • CAS number 159496-16-9; molecular weight 249.16 g/mol.

Catalog No. 50-002-97268 Supplier Medchemexpress LLC Supplier No. HYB1449S1010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine-13C5 is a stable isotope-labeled form of the nucleoside uridine with five 13C-enriched carbon atoms. It is supplied as a research-grade solid intended for use as an internal standard or tracer in metabolic labeling and mass spectrometry applications.

  • Purity 99.9%.
  • Physical form off-white to light brown solid.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for use as an internal standard or tracer in metabolic and mass spectrometry studies.
  • Available in small packaging sizes, including 10 mg.
  • CAS number 159496-16-9; molecular weight 249.16 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.