Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine-1,3-15N2 | 92487-68-8 | 687019 | 98% | 1 MG
SDP

Catalog No. 5000412916
Encompass_Preferred

Uridine-15N2 is the 15N labeled Uridine, used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. It is a stable heavy isotope incorporated into drug molecules for quantitation during drug development. This product appears as a white to light yellow solid and is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Features 98% purity.
  • Exhibits 98.3% isotopic enrichment.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light).

Catalog No. 50-004-12916 Supplier Medchemexpress LLC Supplier No. HYB1449S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uridine-15N2 is the 15N labeled Uridine, used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. It is a stable heavy isotope incorporated into drug molecules for quantitation during drug development. This product appears as a white to light yellow solid and is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Features 98% purity.
  • Exhibits 98.3% isotopic enrichment.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture and light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.