Learn More
Medchemexpress LLC Uridine-1,3-15N2 | 92487-68-8 | 98% | 5 MG
SDP
Catalog No. 5000413525
Encompass_Preferred
Uridine-15N2 is the 15N labeled Uridine, utilized as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes such as hydrogen, carbon, and other elements into drug molecules serves for quantitation during the drug development process. Deuteration has demonstrated the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
Catalog No. 50-004-13525 Supplier Medchemexpress LLC Supplier No. HYB1449S5MG
Uridine-15N2 is the 15N labeled Uridine, utilized as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes such as hydrogen, carbon, and other elements into drug molecules serves for quantitation during the drug development process. Deuteration has demonstrated the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.