Learn More
Medchemexpress LLC Uridine-13C,15N2 | 369656-75-7 | 1 MG
SDP
Catalog No. 5000413711
Encompass_Preferred
Uridine-13C,15N2 is a 13C- and 15N-labeled Uridine. It is intended for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention due to its potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13711 Supplier Medchemexpress LLC Supplier No. HYB1449S91MG
Uridine-13C,15N2 is a 13C- and 15N-labeled Uridine. It is intended for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration has gained attention due to its potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.