Learn More
Medchemexpress LLC Uridine-13C-2 | 478511-14-7 | 99.9% | 245.19 g/mol | C9H12N2O6 | 10 MG
SDP
Catalog No. 5000298037
Encompass_Preferred
Uridine-13C-2 is a 13C-labeled form of the nucleoside uridine, supplied for use as an isotopic tracer or internal standard in analytical and research applications. It is intended for quantitative and structural studies by techniques such as NMR and mass spectrometry.
  • Isotopically labeled at carbon-2 (13C-2).
  • High purity (99.9%) for analytical accuracy.
  • Suitable as an internal standard for NMR, LC-MS, and GC-MS.
  • Provided in small sealed quantities for laboratory use.
  • Intended for research use; follow appropriate handling and safety procedures.
Catalog No. 50-002-98037 Supplier Medchemexpress LLC Supplier No. HYB1449S310MG
Uridine-13C-2 is a 13C-labeled form of the nucleoside uridine, supplied for use as an isotopic tracer or internal standard in analytical and research applications. It is intended for quantitative and structural studies by techniques such as NMR and mass spectrometry.
  • Isotopically labeled at carbon-2 (13C-2).
  • High purity (99.9%) for analytical accuracy.
  • Suitable as an internal standard for NMR, LC-MS, and GC-MS.
  • Provided in small sealed quantities for laboratory use.
  • Intended for research use; follow appropriate handling and safety procedures.