Learn More
Medchemexpress LLC β-Uridine-13C5 | 159496-16-9 | 99.9% | 1 MG
SDP
Catalog No. 5000413699
Encompass_Preferred
Uridine-13C5 (β-Uridine-13C5) is a 13C labeled Uridine (HY-B1449). Uridine (β-Uridine) is a nucleoside compound consisting of uracil and a ribose ring, which are linked by a β-N1- glycosyl bond.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13699 Supplier Medchemexpress LLC Supplier No. HYB1449S101MG
Uridine-13C5 (β-Uridine-13C5) is a 13C labeled Uridine (HY-B1449). Uridine (β-Uridine) is a nucleoside compound consisting of uracil and a ribose ring, which are linked by a β-N1- glycosyl bond.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.