Learn More
Medchemexpress LLC Uridine-13C5 | 159496-16-9 | 99.9% | 5 MG
SDP
Catalog No. 5000412611
Encompass_Preferred
Uridine-13C5 (β-Uridine-13C5) is a 13C labeled Uridine, a nucleoside compound composed of uracil and a ribose ring linked by a β-N1- glycosyl bond. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12611 Supplier Medchemexpress LLC Supplier No. HYB1449S105MG
Uridine-13C5 (β-Uridine-13C5) is a 13C labeled Uridine, a nucleoside compound composed of uracil and a ribose ring linked by a β-N1- glycosyl bond. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS