Learn More
Medchemexpress LLC Uridine-13C9 (β-Uridine-13C9) | 58-96-8 | 96.8% | 1 MG
SDP
Catalog No. 5000413529
Encompass_Preferred
Uridine-13C9 (β-Uridine-13C9) is a 13C9-labeled form of Uridine. Uridine (β-Uridine) is a glycosylated pyrimidine-analog containing uracil attached to a ribose ring (or more specifically, aribofuranose) via a β-N1-glycosidic bond.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13529 Supplier Medchemexpress LLC Supplier No. HYB1449S111MG
Uridine-13C9 (β-Uridine-13C9) is a 13C9-labeled form of Uridine. Uridine (β-Uridine) is a glycosylated pyrimidine-analog containing uracil attached to a ribose ring (or more specifically, aribofuranose) via a β-N1-glycosidic bond.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.