Learn More
Apexbio Technology LLC Uridine 58-96-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063427
Encompass_Preferred
Uridine (CAS 58-96-8) is a nucleoside that serves as a critical biochemical precursor for RNA synthesis through enzymatic polymerization pathways As a pyrimidine nucleoside uridine participates directly in nucleotide metabolism impacting cellular energy processes and nucleotide balance Additionally uridine demonstrates vasodilatory properties exerting effects on vascular tone through mechanisms likely involving purinergic signaling pathways It can also modulate contractility in specific tissue models Due to these characteristics uridine is extensively utilized in biomedical research focusing on RNA metabolism vascular biology and muscle contraction dynamics
Catalog No. 50-000-63427 Supplier Apexbio Technology LLC Supplier No. B14735.1
Uridine (CAS 58-96-8) is a nucleoside that serves as a critical biochemical precursor for RNA synthesis through enzymatic polymerization pathways As a pyrimidine nucleoside uridine participates directly in nucleotide metabolism impacting cellular energy processes and nucleotide balance Additionally uridine demonstrates vasodilatory properties exerting effects on vascular tone through mechanisms likely involving purinergic signaling pathways It can also modulate contractility in specific tissue models Due to these characteristics uridine is extensively utilized in biomedical research focusing on RNA metabolism vascular biology and muscle contraction dynamics